Learn More
ABclonal Technology EcoRV
SDP
Catalog No. 5000136500
Encompass_Preferred
5...GAT/ATC...3 3...CTA/TAG...5 Isoschizomers: Eco32I
Catalog No. 50-001-36500 Supplier ABclonal Technology Supplier No. RK211064000U
5...GAT/ATC...3 3...CTA/TAG...5 Isoschizomers: Eco32I