Learn More
ABclonal Technology HindIII
SDP
Catalog No. 5000135960
Encompass_Preferred
5...A/ A G C T T...3 3...T T C G A/ A...5
Catalog No. 50-001-35960 Supplier ABclonal Technology Supplier No. RK2110810000U
5...A/ A G C T T...3 3...T T C G A/ A...5