Learn More
ABclonal Technology DpnI
SDP
Catalog No. 5000138422
Encompass_Preferred
5...GmA/TC...3 3...CT/mAG...5 Isoschizomers: MalI
Catalog No. 50-001-38422 Supplier ABclonal Technology Supplier No. RK211095000U
5...GmA/TC...3 3...CT/mAG...5 Isoschizomers: MalI