Learn More
ABclonal Technology DpnII
SDP
Catalog No. 5000136526
Encompass_Preferred
5.../G A T C...3 3... C T A G /...5 Isoschizomers: Bsp143I Sau3AI BssMI BstMBI Kzo9I MboI NdeII
Catalog No. 50-001-36526 Supplier ABclonal Technology Supplier No. RK21110500U
5.../G A T C...3 3... C T A G /...5 Isoschizomers: Bsp143I Sau3AI BssMI BstMBI Kzo9I MboI NdeII