Learn More
ABclonal Technology NheI
SDP
Catalog No. 5000135219
Encompass_Preferred
5…G/CTAGC...3 3…CGATC/G...5 Isoschizomers: AsuNHI
Catalog No. 50-001-35219 Supplier ABclonal Technology Supplier No. RK211111000U
5…G/CTAGC...3 3…CGATC/G...5 Isoschizomers: AsuNHI