Learn More
ABclonal Technology NheI
SDP
Catalog No. 5000138428
Encompass_Preferred
5…G/CTAGC...3 3…CGATC/G...5 Isoschizomers: AsuNHI
Catalog No. 50-001-38428 Supplier ABclonal Technology Supplier No. RK211115000U
5…G/CTAGC...3 3…CGATC/G...5 Isoschizomers: AsuNHI