Learn More
ABclonal Technology XbaI
SDP
Catalog No. 5000138423
Encompass_Preferred
5…T/CTAGA...3 3…AGATC/T...5
Catalog No. 50-001-38423 Supplier ABclonal Technology Supplier No. RK2111215000U
5…T/CTAGA...3 3…AGATC/T...5