Learn More
ABclonal Technology XbaI
SDP
Catalog No. 5000135962
Encompass_Preferred
5…T/CTAGA...3 3…AGATC/T...5
Catalog No. 50-001-35962 Supplier ABclonal Technology Supplier No. RK211123000U
5…T/CTAGA...3 3…AGATC/T...5