Learn More
ABclonal Technology SpeI
SDP
Catalog No. 5000136645
Encompass_Preferred
5…A/CTAGT...3 3…TGATC/A...5 Isoschizomers: BcuI AhlI
Catalog No. 50-001-36645 Supplier ABclonal Technology Supplier No. RK211132500U
5…A/CTAGT...3 3…TGATC/A...5 Isoschizomers: BcuI AhlI