Learn More
ABclonal Technology BglI
SDP
Catalog No. 5000135218
Encompass_Preferred
5…GCCNNNN/NGGC...3 3…CGGN/NNNNCCG...5
Catalog No. 50-001-35218 Supplier ABclonal Technology Supplier No. RK211142000U
5…GCCNNNN/NGGC...3 3…CGGN/NNNNCCG...5