Learn More
ABclonal Technology NcoI
SDP
Catalog No. 5000135961
Encompass_Preferred
5…C/CATGG...3 3…GGTAC/C...5 Isoschizomers: Bsp19I
Catalog No. 50-001-35961 Supplier ABclonal Technology Supplier No. RK211151000U
5…C/CATGG...3 3…GGTAC/C...5 Isoschizomers: Bsp19I