Learn More
ABclonal Technology SmaI
SDP
Catalog No. 5000135921
Encompass_Preferred
5…CCC/GGG...3 3…GGG/CCC...5
Catalog No. 50-001-35921 Supplier ABclonal Technology Supplier No. RK2111610000U
5…CCC/GGG...3 3…GGG/CCC...5