Learn More
ABclonal Technology SmaI
SDP
Catalog No. 5000136528
Encompass_Preferred
5…CCC/GGG...3 3…GGG/CCC...5
Catalog No. 50-001-36528 Supplier ABclonal Technology Supplier No. RK211162000U
5…CCC/GGG...3 3…GGG/CCC...5