Learn More
ABclonal Technology StyI
SDP
Catalog No. 5000135965
Encompass_Preferred
5…C/CWWGG...3 3…GGWWC/C...5 Isoschizomers: Eco130I BssT1I EcoT14I ErhI
Catalog No. 50-001-35965 Supplier ABclonal Technology Supplier No. RK2111715000U
5…C/CWWGG...3 3…GGWWC/C...5 Isoschizomers: Eco130I BssT1I EcoT14I ErhI