Learn More
ABclonal Technology MspI
SDP
Catalog No. 5000138430
Encompass_Preferred
5...C/CGG...3 3...GGC/C...5 Isoschizomers: HpaII HapII BsiSI
Catalog No. 50-001-38430 Supplier ABclonal Technology Supplier No. RK211185000U
5...C/CGG...3 3...GGC/C...5 Isoschizomers: HpaII HapII BsiSI