Learn More
ABclonal Technology AvrII
SDP
Catalog No. 5000135920
Encompass_Preferred
5…C/CTAGG...3 3…GGATC/C...5 Isoschizomers: XmaJI AspA2I BlnI
Catalog No. 50-001-35920 Supplier ABclonal Technology Supplier No. RK21119100U
5…C/CTAGG...3 3…GGATC/C...5 Isoschizomers: XmaJI AspA2I BlnI