Learn More
ABclonal Technology SgrAI
SDP
Catalog No. 5000135840
Encompass_Preferred
5…CR/CCGGYG...3 3…GYGGCC/RC...5
Catalog No. 50-001-35840 Supplier ABclonal Technology Supplier No. RK211205000U
5…CR/CCGGYG...3 3…GYGGCC/RC...5