Learn More
ABclonal Technology Acc65I
SDP
Catalog No. 5000136527
Encompass_Preferred
5…G/GTACC...3 3…CCATG/G...5 Isoschizomers: Asp718I
Catalog No. 50-001-36527 Supplier ABclonal Technology Supplier No. RK211212000U
5…G/GTACC...3 3…CCATG/G...5 Isoschizomers: Asp718I