Learn More
ABclonal Technology MluI
SDP
Catalog No. 5000135964
Encompass_Preferred
5…A/CGCGT...3 3…TGCGC/A...5
Catalog No. 50-001-35964 Supplier ABclonal Technology Supplier No. RK211221000U
5…A/CGCGT...3 3…TGCGC/A...5