Learn More
ABclonal Technology SalI
SDP
Catalog No. 5000136647
Encompass_Preferred
5…G/TCGAC...3 3…CAGCT/G...5
Catalog No. 50-001-36647 Supplier ABclonal Technology Supplier No. RK2112310000U
5…G/TCGAC...3 3…CAGCT/G...5