Learn More
ABclonal Technology SalI
SDP
Catalog No. 5000136643
Encompass_Preferred
5…G/TCGAC...3 3…CAGCT/G...5
Catalog No. 50-001-36643 Supplier ABclonal Technology Supplier No. RK211232000U
5…G/TCGAC...3 3…CAGCT/G...5