Learn More
ABclonal Technology AgeI
SDP
Catalog No. 5000135084
Encompass_Preferred
5…A/CCGGT...3 3…TGGCC/A...5 Isoschizomers: BshTI AsiGI CspAI PinAI
Catalog No. 50-001-35084 Supplier ABclonal Technology Supplier No. RK21125300U
5…A/CCGGT...3 3…TGGCC/A...5 Isoschizomers: BshTI AsiGI CspAI PinAI