Learn More
ABclonal Technology SacI
SDP
Supplier:  ABclonal Technology RK2112610000U
Encompass_Preferred
5...G A G C T /C...3 3...C /T C G A G...5 Isoschizomers: Psp124BI, SstI
Catalog No. 50-001-36639