Learn More
ABclonal Technology SacI
SDP
Catalog No. 5000138597
Encompass_Preferred
5...G A G C T /C...3 3...C /T C G A G...5 Isoschizomers: Psp124BI, SstI
Catalog No. 50-001-38597 Supplier ABclonal Technology Supplier No. RK211262000U
5...G A G C T /C...3 3...C /T C G A G...5 Isoschizomers: Psp124BI, SstI