Learn More
ABclonal Technology AscI
SDP
Catalog No. 5000138050
Encompass_Preferred
5...GG/CGCGCC...3 3...CCGCGC/GG...5 Isoschizomers: SgsI PalAI
Catalog No. 50-001-38050 Supplier ABclonal Technology Supplier No. RK21127500U
5...GG/CGCGCC...3 3...CCGCGC/GG...5 Isoschizomers: SgsI PalAI