Learn More
ABclonal Technology SbfI
SDP
Catalog No. 5000138658
Encompass_Preferred
5…C C T G C A/ G G...3 3…G G/ A C G T C C…5 Isoschizomers: SdaI, Sse8387I
Catalog No. 50-001-38658 Supplier ABclonal Technology Supplier No. RK211282500U
5…C C T G C A/ G G...3 3…G G/ A C G T C C…5 Isoschizomers: SdaI, Sse8387I