Learn More
ABclonal Technology SbfI
SDP
Supplier:  ABclonal Technology RK21128500U
Encompass_Preferred
5…C C T G C A/ G G...3 3…G G/ A C G T C C…5 Isoschizomers: SdaI, Sse8387I
Catalog No. 50-001-35117