Learn More
ABclonal Technology BbsI
SDP
Catalog No. 5000135308
Encompass_Preferred
5...G A A G A C (N)2 ...3 3...C T T C T G (N)6 ...5 Isoschizomers: BpiI, BstV2I
Catalog No. 50-001-35308 Supplier ABclonal Technology Supplier No. RK21130500U
5...G A A G A C (N)2 ...3 3...C T T C T G (N)6 ...5 Isoschizomers: BpiI, BstV2I