Learn More
ABclonal Technology BstEII
SDP
Catalog No. 5000136516
Encompass_Preferred
5...G/ G T N A C C...3 3...C C A N T G/ G...5 Isoschizomers: Eco91I BstPI EcoO65I PspEI
Catalog No. 50-001-36516 Supplier ABclonal Technology Supplier No. RK211521000U
5...G/ G T N A C C...3 3...C C A N T G/ G...5 Isoschizomers: Eco91I BstPI EcoO65I PspEI