Learn More
ABclonal Technology ClaI
SDP
Catalog No. 5000138028
Encompass_Preferred
5...A T/ C G A T...3 3...T A G C/ T A...5 Isoschizomers: Bsu15I Bsa29I BseCI BshVI BspDI BsuTUI
Catalog No. 50-001-38028 Supplier ABclonal Technology Supplier No. RK21153500U
5...A T/ C G A T...3 3...T A G C/ T A...5 Isoschizomers: Bsu15I Bsa29I BseCI BshVI BspDI BsuTUI