Learn More
ABclonal Technology HpaI
SDP
Catalog No. 5000136551
Encompass_Preferred
5...G T T/ A A C...3 3...C A A/ T T G...5 Isoschizomers: KspAI
Catalog No. 50-001-36551 Supplier ABclonal Technology Supplier No. RK21155500U
5...G T T/ A A C...3 3...C A A/ T T G...5 Isoschizomers: KspAI