Learn More
ABclonal Technology NdeI
SDP
Catalog No. 5000135015
Encompass_Preferred
5...C A/ T A T G...3 3...G T A T/ A C...5 Isoschizomers: FauNDI
Catalog No. 50-001-35015 Supplier ABclonal Technology Supplier No. RK211574000U
5...C A/ T A T G...3 3...G T A T/ A C...5 Isoschizomers: FauNDI