Learn More
ABclonal Technology PstI
SDP
Catalog No. 5000138522
Encompass_Preferred
5...C T G C A/ G...3 3...G/ A C G T C...5 Isoschizomers: BspMAI
Catalog No. 50-001-38522 Supplier ABclonal Technology Supplier No. RK2115910000U
5...C T G C A/ G...3 3...G/ A C G T C...5 Isoschizomers: BspMAI