Learn More
ABclonal Technology SphI
SDP
Catalog No. 5000138520
Encompass_Preferred
5...G C A T G/ C...3 3...C/ G T A C G...5 Isoschizomers: PaeI
Catalog No. 50-001-38520 Supplier ABclonal Technology Supplier No. RK211601000U
5...G C A T G/ C...3 3...C/ G T A C G...5 Isoschizomers: PaeI