Learn More
ABclonal Technology TaqI
SDP
Catalog No. 5000135118
Encompass_Preferred
5...T/ C G A...3 3...A G C/ T...5
Catalog No. 50-001-35118 Supplier ABclonal Technology Supplier No. RK211634000U
5...T/ C G A...3 3...A G C/ T...5