Learn More
ABclonal Technology BglII
SDP
Catalog No. 5000138030
Encompass_Preferred
5...A/ G A T C T...3 3...T C T A G/ A...5
Catalog No. 50-001-38030 Supplier ABclonal Technology Supplier No. RK211652000U
5...A/ G A T C T...3 3...T C T A G/ A...5