Learn More
ABclonal Technology PvuII
SDP
Catalog No. 5000138521
Encompass_Preferred
5...C A G/ C T G...3 3...G T C/ G A C...5
Catalog No. 50-001-38521 Supplier ABclonal Technology Supplier No. RK211684000U
5...C A G/ C T G...3 3...G T C/ G A C...5