Learn More
ABclonal Technology ApaLI
SDP
Catalog No. 5000137759
Encompass_Preferred
5...G/TGCAC...3 3...CACGT/G...5 Isoschizomers: Alw44I VneI
Catalog No. 50-001-37759 Supplier ABclonal Technology Supplier No. RK211702000U
5...G/TGCAC...3 3...CACGT/G...5 Isoschizomers: Alw44I VneI