Learn More
ABclonal Technology PacI
SDP
Catalog No. 5000138029
Encompass_Preferred
5…TTAAT/TAA...3 3…AAT/TAATT...5
Catalog No. 50-001-38029 Supplier ABclonal Technology Supplier No. RK21171250U
5…TTAAT/TAA...3 3…AAT/TAATT...5