Learn More
ABclonal Technology SacII
SDP
Catalog No. 5000136552
Encompass_Preferred
5...C C G C/ G G...3 3...G G/ C G C C...5 Isoschizomers: Cfr42I KspI Sfr303I SgrBI
Catalog No. 50-001-36552 Supplier ABclonal Technology Supplier No. RK211721000U
5...C C G C/ G G...3 3...G G/ C G C C...5 Isoschizomers: Cfr42I KspI Sfr303I SgrBI