Learn More
ABclonal Technology BsmBI
SDP
Catalog No. 5000138523
Encompass_Preferred
5...C G T C T C(N)1...3 3...G C A G A G(N)5...5 Isoschizomers: Esp3I
Catalog No. 50-001-38523 Supplier ABclonal Technology Supplier No. RK211731000U
5...C G T C T C(N)1...3 3...G C A G A G(N)5...5 Isoschizomers: Esp3I