Learn More
ABclonal Technology BstBI
SDP
Catalog No. 5000137760
Encompass_Preferred
5...T T/ C G A A...3 3...A A G C/ T T...5 Isoschizomers: Bsp119I AsuII Bpu14I BspT104I NspV SfuI
Catalog No. 50-001-37760 Supplier ABclonal Technology Supplier No. RK211742000U
5...T T/ C G A A...3 3...A A G C/ T T...5 Isoschizomers: Bsp119I AsuII Bpu14I BspT104I NspV SfuI