Learn More
ABclonal Technology NsiI
SDP
Catalog No. 5000136611
Encompass_Preferred
5...A/ T G C A T...3 3...T A C G T/ A...5 Isoschizomers: Mph1103I EcoT22I Zsp2I
Catalog No. 50-001-36611 Supplier ABclonal Technology Supplier No. RK21175500U
5...A/ T G C A T...3 3...T A C G T/ A...5 Isoschizomers: Mph1103I EcoT22I Zsp2I