Learn More
ABclonal Technology BspQI
SDP
Catalog No. 5000135145
Encompass_Preferred
5...G C T C T T C (N)1...3 3...C G A G A A G (N)4...5 Isoschizomers: LguI, SapI, PciSI
Catalog No. 50-001-35145 Supplier ABclonal Technology Supplier No. RK21176500U
5...G C T C T T C (N)1...3 3...C G A G A A G (N)4...5 Isoschizomers: LguI, SapI, PciSI