Learn More
ABclonal Technology BsrGI
SDP
Catalog No. 5000138636
Encompass_Preferred
5...T/ G T A C A...3 3...A C A T G/ T...5 Isoschizomers: Bsp1407I BstAUI
Catalog No. 50-001-38636 Supplier ABclonal Technology Supplier No. RK21177600U
5...T/ G T A C A...3 3...A C A T G/ T...5 Isoschizomers: Bsp1407I BstAUI