Learn More
ABclonal Technology XcmI
SDP
Catalog No. 5000137776
Encompass_Preferred
5...C C A N N N N N/ N N N N T G G...3 3...G G T N N N N/ N N N N N A C C...5
Catalog No. 50-001-37776 Supplier ABclonal Technology Supplier No. RK21179300U
5...C C A N N N N N/ N N N N T G G...3 3...G G T N N N N/ N N N N N A C C...5