Learn More
ABclonal Technology HpaII
SDP
Catalog No. 5000135147
Encompass_Preferred
5...C/CGG...3 3...GGC/C...5 Isoschizomers: MspI HapII BsiSI
Catalog No. 50-001-35147 Supplier ABclonal Technology Supplier No. RK211802000U
5...C/CGG...3 3...GGC/C...5 Isoschizomers: MspI HapII BsiSI