Learn More
ABclonal Technology ScaI
SDP
Catalog No. 5000135148
Encompass_Preferred
5... A G T /A C T...3 3... T C A /T G A...5 Isoschizomers: BmcAI, ZrmI
Catalog No. 50-001-35148 Supplier ABclonal Technology Supplier No. RK211812000U
5... A G T /A C T...3 3... T C A /T G A...5 Isoschizomers: BmcAI, ZrmI