Learn More
ABclonal Technology Genious 2X SYBR Green Fast qPCR Mix
SDP
Supplier:  ABclonal Technology RK2120425ML
Encompass_Preferred
Real-time Quantitative PCR (qPCR) is a powerful technique in detecting initial DNA input in a PCR reaction by fluorescence signal accumulation. ABclonal Genious 2X SYBR Green Fast qPCR Mix is a fluorescence reagent for qPCR reactions using the SYBR(R) Green I. This product provides real-time data on DNA amplification during PCR by performing quantitative fluorescence signal detection on SYBR(R) / FAM channels. This product uses the antibody-based hot-start Taq enzyme for amplification, which greatly improves the specificity of the product while ensuring the amplification effect. At the same time, by optimizing the qPCR Mix Buffer system, the product is suitable for multiple species, providing a powerful tool for multidisciplinary experimental needs. This product is a 2X pre-mix enzyme, contains all the components required for qPCR except primers and templates, providing great convenience for experimental manipulation.
Catalog No. 50-001-35149