Learn More
ABclonal Technology BrightCycle Universal SYBR Green qPCR Mix with UDG
SDP
Catalog No. 5000135047
Encompass_Preferred
This product is a universal SYBR Green reagent that uses a specially designed reference dye (ROX), has a more sensitive resolution, and can be applied to all types of fluorescence quantitative PCR instruments on the market (including High ROX, Low ROX, and No ROX required instruments). This reagent introduces a dUTP/UDG anti fouling system. UDG can quickly degrade pollutants containing U at room temperature, and UDG quickly deactivates when pre denatured at 95 ℃, without affecting the efficiency and sensitivity of qPCR. This product uses hot start Taq DNA polymerase for amplification, which greatly improves the specificity of the product while ensuring the amplification effect.
Catalog No. 50-001-35047 Supplier ABclonal Technology Supplier No. RK2121925ML
This product is a universal SYBR Green reagent that uses a specially designed reference dye (ROX), has a more sensitive resolution, and can be applied to all types of fluorescence quantitative PCR instruments on the market (including High ROX, Low ROX, and No ROX required instruments). This reagent introduces a dUTP/UDG anti fouling system. UDG can quickly degrade pollutants containing U at room temperature, and UDG quickly deactivates when pre denatured at 95 ℃, without affecting the efficiency and sensitivity of qPCR. This product uses hot start Taq DNA polymerase for amplification, which greatly improves the specificity of the product while ensuring the amplification effect.